missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCL1A Antibody (1-21), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-43253PE
This item is not returnable.
View return policy
Description
TCL1A Monoclonal specifically detects TCL1A in Human samples. It is validated for Flow Cytometry.
Specifications
| TCL1A | |
| Monoclonal | |
| PE | |
| Oncogene TCL1, Oncogene TCL-1, Protein p14 TCL1, T-cell leukemia/lymphoma 1A, T-cell lymphoma-1, T-cell lymphoma-1A, TCL1T-cell leukemia/lymphoma protein 1A | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 8115 | |
| Store at 4C in the dark. |
| Flow Cytometry | |
| 43121 | |
| Flow Cytometry | |
| TCL1A | |
| The immunogen for this antibody was TCL1. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction