Learn More
Invitrogen™ TCIRG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590425
Description
Immunogen sequence: MGSMFRSEEV ALVQLFLPTA AAYTCVSRLG ELGLVEFRDL NASVSAFQRR FVVDVRRCEE LEKTFTFLQE EVRRAGLVLP PPKGRLPAPP PRDLLRIQEE TERLAQELRD VRGNQQALRA QLHQLQLHAA; Positive Samples: U2OS, A-549, BxPC-3, U-937, HL-60, Mouse thymus, Mouse skeletal muscle, Rat thymus, Rat skeletal muscle; Cellular Location: Membrane, Multi-pass membrane protein.
The T-cell immune regulator 1 (TCIRG1) is the a3 subunit of the vacuolar-type proton transporting ATPase (V-ATPase), a proton pump in late endosomes and lysosomes that functions in the luminal acidification of these organelles. Genetic defects in the gene encoding this protein a responsible for a severe form of autosomal recessive osteopetrosis, a condition that leads to increased bone density, decreased bone strength, and inflammation in bone tissues. TCIRG1 is also required for the normal secretion of insulin and the bacteria-killing function of macrophages.
Specifications
| TCIRG1 | |
| Polyclonal | |
| Unconjugated | |
| TCIRG1 | |
| a3; ATP6a3; Atp6i; ATP6N1C; ATP6V0A3; ATPase, H+ transporting, 116kD; ATPase, H+ transporting, lysosomal I; ATPase, H+ transporting, lysosomal V0 protein a; oc; OC116; OC-116; OC-116 kDa; OC-116kDa; OPTB1; osteoclastic proton pump; osteoclastic proton pump 116 kDa subunit; osteosclerotic; specific 116-kDa vacuolar proton pump subunit; Stv1; T cell immune response cDNA7 protein; T cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein A3; T-cell immune regulator 1; T-cell immune regulator 1, ATPase H+ transporting V0 subunit a3; T-cell immune response cDNA 7; T-cell immune response cDNA7 protein; T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein a; T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein A3; T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 subunit A3; TCIRG1; TIRC7; vacuolar proton translocating ATPase 116 kDa subunit A; Vacuolar proton translocating ATPase 116 kDa subunit a isoform 3; V-ATPase 116 kDa isoform a3; V-ATPase 116-kDa; V-ATPase a3; v-H+ATPase subunit a3; Vph1; V-type proton ATPase 116 kDa subunit a; V-type proton ATPase 116 kDa subunit a isoform 3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10312, 27060, 293650 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2.22 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q13488 | |
| TCIRG1 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.