missing translation for 'onlineSavingsMsg'
Learn More

TCIRG1 Antibody, Novus Biologicals™

Product Code. 18337239 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18337239 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18337239 Supplier Novus Biologicals Supplier No. H00010312B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

TCIRG1 Polyclonal antibody specifically detects TCIRG1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCIRG1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_006044.1
Gene Alias a3, Atp6i, ATP6N1Cspecific 116-kDa vacuolar proton pump subunit, ATP6V0A3T-cell immune response cDNA 7, OC-116, OC-116 kDa, OC-116kDa, OC116Vph1, OPTB1, Osteoclastic proton pump 116 kDa subunit, Stv1, T-cell immune regulator 1, T-cell immune response cDNA7 protein, T-cell, immune regulator 1, T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein a, T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein A3, T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein aisoform 3, T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 subunit A3, TIRC7ATPase, H+ transporting, 116kD, vacuolar proton translocating ATPase 116 kDa subunit A, Vacuolar proton translocating ATPase 116 kDa subunit a isoform 3, V-ATPase 116 kDa isoform a3, V-ATPase 116-kDa, V-type proton ATPase 116 kDa subunit a, V-type proton ATPase 116 kDa subunit a isoform 3
Host Species Mouse
Immunogen TCIRG1 (NP_006044.1, 1 a.a. - 614 a.a.) full-length human protein. MTFLISYWGEQIGQKIRKITDCFHCHVFPFLQQEEARLGALQQLQQQSQELQEVLGETERFLSQVLGRVLQLLPPGQVQVHKMKAVYLALNQCSVSTTHKCLIAEAWCSVRDLPALQEALRDSSMEEGVSAVAHRIPCRDMPPTLIRTNRFTASFQGIVDAYGVGRYQEVNPAPYTIITFPFLFAVMFGDVGHGLLMFLFALAMVLAENRPAVKAAQNEIWQTFFRGRYLLLLMGLFSIYTGFIYNECFSRATSIFPSGWSVAAMANQSGWSDAFLAQHTMLTLDPNVTGVFLGPYPFGIDPIWSLAANHLSFLNSFKMKMSVILGVVHMAFGVVLGVFNHVHFGQRHRLLLETLPELTFLLGLFGYLVFLVIYKWLCVWAARAASAPSILIHFINMFLFSHSPSNRLLYPRQEVVQATLVVLALAMVPILLLGTPLHLLHRHRRRLRRRPADRQEENKAGLLDLPDASVNGWSSDEEKAGGLDDEEEAELVPSEVLMHQAIHTIEFCLGCVSNTASYLRLWALSLAHAQLSEVLWAMVMRIGLGLGREVGVAAVVLVPIFAAFAVMTVAILLVMEGLSAFLHALRLHWVEFQNKFYSGTGYKLSPFTFAATDD
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10312
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.