missing translation for 'onlineSavingsMsg'
Learn More

TCIRG1 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18430136 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430136 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430136 Supplier Novus Biologicals Supplier No. NBP281936AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TCIRG1 Polyclonal antibody specifically detects TCIRG1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCIRG1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias a3, Atp6i, ATP6N1Cspecific 116-kDa vacuolar proton pump subunit, ATP6V0A3T-cell immune response cDNA 7, OC-116, OC-116 kDa, OC-116kDa, OC116Vph1, OPTB1, Osteoclastic proton pump 116 kDa subunit, Stv1, T-cell immune regulator 1, T-cell immune response cDNA7 protein, T-cell, immune regulator 1, T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein a, T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein A3, T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein aisoform 3, T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 subunit A3, TIRC7ATPase, H+ transporting, 116kD, vacuolar proton translocating ATPase 116 kDa subunit A, Vacuolar proton translocating ATPase 116 kDa subunit a isoform 3, V-ATPase 116 kDa isoform a3, V-ATPase 116-kDa, V-type proton ATPase 116 kDa subunit a, V-type proton ATPase 116 kDa subunit a isoform 3
Host Species Rabbit
Immunogen TCIRG1 antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human TCIRG1. The immunogen is located within amino acids 70 - 120 of TCIRG1. Amino Acid Squence: PPKGRLPAPPPRDLLRIQE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10312
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.