missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TCID
Description
TCID is a selective ubiquitin C-terminal hydrolase-L3 (UCH-L3) inhibitor (IC50 = 0.6 μM). Exhibits >100-fold selectivity for UCH-L3 over UCH-L1. Diminishes glycine transporter GlyT2 ubiquitination in brain stem and spinal cord primary neurons.
Specifications
Specifications
| Purity | >99% |
| Quantity | 10 mg |
| Product Type | TCID |
| Molecular Weight (g/mol) | 283.92 |
| Inhibitors | TCID |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?