missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCHP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563974
This item is not returnable.
View return policy
Description
Immunogen sequence: LREEAKEMWE KREAEWARER SARDRLMSEV LTGRQQQIQE KIEQNRRAQE ESLKHREQLI RNLEEVRELA RREK Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.
TCHP may act as a 'capping' or 'branching' protein for keratin filaments in the cell periphery. This protein may regulate K8/K18 filament and desmosome organization mainly at the apical or peripheral regions of simple epithelial cells.
Specifications
| TCHP | |
| Polyclonal | |
| Unconjugated | |
| TCHP | |
| A930031F18Rik; AV279199; Mitochondrial protein with oncostatic activity; mitostatin; Protein TCHP; Tchp; TpMs; trichoplein keratin filament binding; trichoplein keratin filament-binding protein; trichoplein, keratin filament binding; tumor suppressor protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84260 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BT92 | |
| TCHP | |
| Recombinant protein corresponding to Human TCHP. Recombinant protein control fragment (Product #RP-103948). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction