missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TCH-165
GREENER_CHOICE

Product Code. p-7223003
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18765466 10 mg 10mg
18746176 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18765466 Supplier Tocris Bioscience™ Supplier No. 7489/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Enhancer of 20S proteasome assembly and 20S-mediated protein degradation

TCH-165 is an enhancer of 20S proteasome assembly. TCH-165 regulates the dynamic equilibrium between the 20S and 26S proteasome complexes, favoring 20S-mediated protein degradation. TCH-165 enhances the proteolytic activity of the 20S proteasome by increasing substrate accessibility to the 20S catalytic chamber through 20S gate opening. TCH-165 enhances 20S-mediated degradation of IDPs, α-syn, and tau in vitro, and does not induce the degradation of structured proteins such as GAPDH. TCH-165 enhances c-MYC degradation by the 20S proteasome, which reduces c-MYC levels in vivo, and is effective in bortezomib (Cat. No. 7282) resistant cells. TCH-165 down-regulates c-MYC target genes and inhibits cancer cell proliferation and tumor growth in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl (4 R,5 R)-rel-1-benzyl-5-(4-(benzylamino)phenyl)-2-(4-methoxyphenyl)-4-phenyl-4,5-dihydro-1 H-imidazole-4-carboxylate
CAS 1446350-60-2
Quantity 10 mg
Target Proteasome Modulators
Molecular Formula C39H37N3O3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.