missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCFL5 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518588
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
TCFL5 is a new bHLH transcription factor that negatively regulates upstream transcription factor-dependent transcription.
Specifications
| TCFL5 | |
| Polyclonal | |
| Unconjugated | |
| TCFL5 | |
| AV260129; bHLHe82; CHA; Cha transcription factor; E2BP1; E2BP-1; factor in the germline beta; FIG beta; Figlb; HPV-16 E2-binding protein 1; LOW QUALITY PROTEIN: transcription factor-like 5 protein; MGC46135; RGD1305899; SOSF1; TCFL5; transcription factor like 5; transcription factor-like 5 (basic helix-loop-helix); transcription factor-like 5 protein | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 10732 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9UL49 | |
| TCFL5 | |
| Synthetic peptide sequence (KYIQERHGDSLKKE) corresponding to the internal amino acids of TCFL5 (aa 448-462). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction