missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCFL5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599728
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TCFL5.
TCFL5 is a new bHLH transcription factor that negatively regulates upstream transcription factor-dependent transcription.
Specifications
| TCFL5 | |
| Polyclonal | |
| Unconjugated | |
| TCFL5 | |
| AV260129; bHLHe82; CHA; Cha transcription factor; E2BP1; E2BP-1; factor in the germline beta; FIG beta; Figlb; HPV-16 E2-binding protein 1; LOW QUALITY PROTEIN: transcription factor-like 5 protein; MGC46135; RGD1305899; SOSF1; TCFL5; transcription factor like 5; transcription factor-like 5 (basic helix-loop-helix); transcription factor-like 5 protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10732, 277353 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UL49 | |
| TCFL5 | |
| A synthesized peptide derived from human TCFL5(Accession Q9UL49), corresponding to amino acid residues P428-R478. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction