missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCF9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA537072
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 89 and 120 kDa. Purity is >95% by SDS-PAGE.
TCF-9 (transcription factor 9), also known as C2orf3 (chromosome 2 open reading frame 3), GCF (GC-rich sequence DNA-binding factor) or DNABF, is a 781 amino acid nuclear protein that belongs to the GCF family. Widely expressed, GCF binds the GC-rich sequences of beta-Actin, EGFR and calcium-dependent protease (CANP) promoters. GCF contains multiple phosphoserine and phosphothreonine residues, and two GCF isoforms are produced due to alternative splicing events. GCF is considered a candidate for susceptibility to dyslexia (DYX3) as both genes reside in close proximity on human chromosome 2. Chromosome 2 is the second largest human chromosome and consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome.
Specifications
| TCF9 | |
| Polyclonal | |
| Unconjugated | |
| GCFC2 | |
| A130099G21; AW146020; C2orf3; DNABF; GC binding factor; GC bindng factor; GCF; GCF2; GCFC2; GC-rich sequence DNA binding factor 2; GC-rich sequence DNA-binding factor; GC-rich sequence DNA-binding factor 2; RGD1304792; TCF9; TCF-9; Transcription factor 9; transcription factor 9 (binds GC-rich sequences) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 312474, 330361, 6936 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide | |
| P16383, Q8BKT3 | |
| GCFC2 | |
| Synthetic peptide corresponding to amino acids 193-238 of human TCF-9. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction