missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF7L2 Antibody (CL11547) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21197JF669
This item is not returnable.
View return policy
Description
TCF7L2 Monoclonal antibody specifically detects TCF7L2 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| TCF7L2 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| HMG box transcription factor 4, hTCF-4, T-cell factor 4, T-cell factor-4 variant A, T-cell factor-4 variant B, T-cell factor-4 variant C, T-cell factor-4 variant D, T-cell factor-4 variant E, T-cell factor-4 variant H, T-cell factor-4 variant I, T-cell factor-4 variant J, T-cell factor-4 variant K, T-cell factor-4 variant L, T-cell factor-4 variant M, T-cell factor-4 variant X2, T-cell-specific transcription factor 4, TCF-4T-cell factor-4 variant F, TCF4T-cell factor-4 variant G, transcription factor 7-like 2, transcription factor 7-like 2 (T-cell specific, HMG-box) | |
| This antibody was developed using a synthetic peptide derived from Q9NQB0, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Signal Transduction, Wnt Signaling Pathway | |
| 6934 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL11547 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto