missing translation for 'onlineSavingsMsg'
Learn More

TCF7L1/TCF3 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18398031 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18398031 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18398031 Supplier Novus Biologicals Supplier No. NBP298907G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TCF7L1/TCF3 Polyclonal specifically detects TCF7L1/TCF3 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCF7L1/TCF3
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias HMG box transcription factor 3, TCF3, TCF-3, transcription factor 7-like 1, transcription factor 7-like 1 (T-cell specific, HMG-box)
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human TCF7L1/TCF3.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Signal Transduction, Stem Cell Markers, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 83439
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.