missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF7/TCF1 Antibody (812145) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB8224JF635
This item is not returnable.
View return policy
Description
TCF7/TCF1 Monoclonal antibody specifically detects TCF7/TCF1 in Mouse samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| TCF7/TCF1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| FLJ36364, MGC47735, T-cell factor 1, T-cell specific, TCF1, TCF-7, transcription factor 7 (T-cell specific, HMG-box) | |
| E. coli-derived recombinant mouse TCF7/TCF1, Gly61-Gly116, Accession # Q00417 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 812145 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Signal Transduction, Wnt Signaling Pathway | |
| 6932 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction