missing translation for 'onlineSavingsMsg'
Learn More

TCF7/TCF1 Antibody (1B3), Novus Biologicals™

Product Code. 18389829 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389829 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389829 Supplier Novus Biologicals Supplier No. H00006932M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TCF7/TCF1 Monoclonal antibody specifically detects TCF7/TCF1 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCF7/TCF1
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1B3
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_003193
Gene Alias FLJ36364, MGC47735, T-cell factor 1, T-cell specific, TCF1, TCF-7, transcription factor 7 (T-cell specific, HMG-box)
Host Species Mouse
Immunogen TCF7 (NP_003193, 287 a.a. ~ 384 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VIAECTLKESAAINQILGRRWHALSREEQAKYYELARKERQLHMQLYPGWSARDNYGKKKRRSREKHQESTTGGKRNAFGTYPEKAAAPAPFLPMTVL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 6932
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.