missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TCF7 |
|---|---|
| Concentration | 0.15 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16893114
|
Proteintech
14464-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16883114
|
Proteintech
14464-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The T cell specific transcription factor TCF7 activates genes involved in immune regulation and thymocyte differentiation, and is a candidate locus for genetic susceptibility to type 1 diabetes.Specifications
| TCF7 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| P36402, Q00417 | |
| 21414, 6932 | |
| TCF7 Fusion Protein Ag5792 | |
| Primary | |
| -20°C | |
| TCF7 |
| 0.15 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| T cell factor 1, TCF 1, TCF 7, TCF1, TCF7, Transcription factor 7 | |
| TCF7 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title