missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF4 Antibody (7B1B3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£163.00 - £437.00
Specifications
| Antigen | TCF4 |
|---|---|
| Clone | 7B1B3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18050504
|
Novus Biologicals
NBP2-61897 |
0.1 mL |
£437.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633568
|
Novus Biologicals
NBP2-61897-0.025ml |
0.025 mL |
£163.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCF4 Monoclonal specifically detects TCF4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| TCF4 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| BHLHB19, bHLHb19E2-2, Class B basic helix-loop-helix protein 19, Immunoglobulin transcription factor 2, ITF-2, ITF2PTHS, MGC149724, SEF-2, SEF2-1, SEF2-1A, SEF2-1B, SEF2MGC149723, SL3-3 enhancer factor 2, TCF-4, transcription factor 4 | |
| TCF4 | |
| IgG2b | |
| Protein G purified |
| 7B1B3 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| Transcription Factors and Regulators | |
| 6925 | |
| Purified recombinant fragment of human TCF4 (AA: 518-667) expressed in E. Coli. | |
| Primary | |
| 147 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title