missing translation for 'onlineSavingsMsg'
Learn More

TCF4 Antibody (3H4), Novus Biologicals™

Product Code. 18378969 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18378969 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18378969 Supplier Novus Biologicals Supplier No. H00006925M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TCF4 Monoclonal antibody specifically detects TCF4 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCF4
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 3H4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. BC031056
Gene Alias BHLHB19, bHLHb19E2-2, Class B basic helix-loop-helix protein 19, Immunoglobulin transcription factor 2, ITF-2, ITF2PTHS, MGC149724, SEF-2, SEF2-1, SEF2-1A, SEF2-1B, SEF2MGC149723, SL3-3 enhancer factor 2, TCF-4, transcription factor 4
Host Species Mouse
Immunogen TCF4 (AAH31056.1, 1 a.a. ~ 365 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MHHQQRMAALGTDKELSDLLDFSAMFSPPVSSGKNGPTSLASGHFTGSNVEDRSSSGSWGNGGHPSPSRNYGDGTPYDHTTSRDLGSHDNLSPPFVNSRIQSKTERGSYSSYGRESNLQGCHQQSLLGGDMDMGNPGTLSPTKPGSQYYQYSSNNPRRRPLHSSAMEVQTKKVRKVPPGLPSSVYAPSASTADYNRDSPGYPSSKPATSTFPSSFFMQDGHHSSDPWSSSSGMNQPGYAGMLGNSSHIPQSSSYCSLHPHERLSYPSHSSADINSSLPPMSTFHRSGTNHYSTSSCTPPANGTDSIMANRGSGAAGSSQTGDALGKALASIYSPDHTNNSFSSNPSTPVGSPPSLSAGTAVWSRN
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6925
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.