missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCF21 Recombinant Rabbit Monoclonal Antibody (24GB2410)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557291
This item is not returnable.
View return policy
Description
TCF21 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The basic helix-loop-helix transcription factor TCF21/Pod1 (also called capsulin or epicardin) is involved in kidney, lung and spleen organogenesis. It is also essential for normal development of the testes and ovaries. TCF21/Pod1 is involved in the transcriptional repression of steroidogenic factor 1 (Sf1/Nr5a1/Ad4BP), an orphan nuclear receptor that regulates the expression of multiple genes (including Scc) that mediate sexual differentiation.
Specifications
| TCF21 | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43680 | |
| TCF21 | |
| A synthesized peptide derived from human TCF21 (aa 110-179). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2410 | |
| Unconjugated | |
| TCF21 | |
| BHLHA23; capsulin; class A basic helix-loop-helix protein 23; epc; Epicardin; hypothetical protein LOC558148; Msf-1; MyoRa2; Pod1; pod-1; podocyte-expressed 1; TCF21; TCF-21; Transcription factor 21; zgc:123313 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6943 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction