missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCF21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572640
This item is not returnable.
View return policy
Description
TCF21 Polyclonal Antibody for Western Blot
The basic helix-loop-helix transcription factor TCF21/Pod1 (also called capsulin or epicardin) is involved in kidney, lung and spleen organogenesis. It is also essential for normal development of the testes and ovaries. TCF21/Pod1 is involved in the transcriptional repression of steroidogenic factor 1 (Sf1/Nr5a1/Ad4BP), an orphan nuclear receptor that regulates the expression of multiple genes (including Scc) that mediate sexual differentiation.
Specifications
| TCF21 | |
| Polyclonal | |
| Unconjugated | |
| TCF21 | |
| BHLHA23; capsulin; class A basic helix-loop-helix protein 23; epc; Epicardin; hypothetical protein LOC558148; Msf-1; MyoRa2; Pod1; pod-1; podocyte-expressed 1; TCF21; TCF-21; Transcription factor 21; zgc:123313 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 558148 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q32PV5 | |
| TCF21 | |
| KLH conjugated synthetic peptide between 5-35 amino acids from the N-terminal region of DANRE tcf21. | |
| 400 μL | |
| Primary | |
| Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction