Learn More
TCF12 Mouse anti-Human, Unconjugated, Clone: 1B8D4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67523-1-IG-150UL
This item is not returnable.
View return policy
Description
The TCF12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TCF12. This antibody recognizes human, mouse, rat antigen. The TCF12 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TCF12 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, ELISA | |
| 1B8D4 | |
| Unconjugated | |
| 150 μL | |
| 6938 | |
| IgG2a |