missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF12/HEB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14419-1-AP-150UL
This item is not returnable.
View return policy
Description
TCF12, also known as HTF4, is a member of the basic helix-loop-helix (bHLH) E-protein family that recognizes the consensus binding site (E-box) CANNTG. TCF12 is expressed in many tissues, among them skeletal muscle, thymus, B- and T-cells, and may participate in regulating lineage-specific gene expression through the formation of heterodimers with other bHLH E-proteins. TCF12, in combination with E2A, is required to block thymocyte proliferation prior to pre-TCR expression and is critical for proper T cell differentiation. Recent reports have shown that TCF12 is also a critical factor required for the development of invariant natural killer T cells.Specifications
| TCF12/HEB | |
| Polyclonal | |
| Unconjugated | |
| TCF12 | |
| bHLHb20, DNA binding protein HTF4, E box binding protein, HEB, HsT17266, HTF4, TCF 12, TCF12, transcription factor 12, Transcription factor HTF 4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21406, 25720, 6938 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P51514, Q61286, Q99081 | |
| TCF12 | |
| TCF12/HEB Fusion Protein Ag5765 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction