missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF-7/TCF-1 Mouse anti-Mouse, Human, BV421, Clone: S33-966, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566692
This item is not returnable.
View return policy
Description
TCF-7/TCF-1 Monoclonal antibody specifically detects TCF-7/TCF-1 in Mouse, Human samples. It is validated for Flow Cytometry
Specifications
| TCF-7/TCF-1 | |
| Monoclonal | |
| Brilliant Violet 421 | |
| TCF7; Transcription factor 7; TCF-1; TCF1; T-cell factor 1 | |
| Mouse TCF-7 Recombinant Protein | |
| RUO | |
| Mouse, Human | |
| IgG1 κ |
| Flow Cytometry | |
| S33-966 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction