missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF-12/HTF4 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2338C
This item is not returnable.
View return policy
Description
TCF-12/HTF4 Polyclonal antibody specifically detects TCF-12/HTF4 in Human samples. It is validated for Western Blot
Specifications
| TCF-12/HTF4 | |
| Polyclonal | |
| Western Blot | |
| BHLHB20, bHLHb20E-box-binding protein, Class B basic helix-loop-helix protein 20, HEBDNA-binding protein HTF4, helix-loop-helix transcription factor 4, HsT17266, HTF4TCF-12, transcription factor 12, Transcription factor HTF-4 | |
| The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human Transcription Factor 12 using the numbering given in entry NP_996919.1 (GeneID 6938). | |
| 0.1 mL | |
| Immunology | |
| 6938 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction