missing translation for 'onlineSavingsMsg'
Learn More

TCF-12/HTF4 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18411316 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411316 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411316 Supplier Novus Biologicals Supplier No. NBP176304AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TCF-12/HTF4 Polyclonal antibody specifically detects TCF-12/HTF4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCF-12/HTF4
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias BHLHB20, bHLHb20E-box-binding protein, Class B basic helix-loop-helix protein 20, HEBDNA-binding protein HTF4, helix-loop-helix transcription factor 4, HsT17266, HTF4TCF-12, transcription factor 12, Transcription factor HTF-4
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human TCF12. The immunogen is located within amino acids 50 - 100 of TCF12. Amino Acid Squence: QPSPSYDSSRGFTDSPHYS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 6938
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.