missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TCF-12/HTF4 Polyclonal antibody specifically detects TCF-12/HTF4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | TCF-12/HTF4 |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | BHLHB20, bHLHb20E-box-binding protein, Class B basic helix-loop-helix protein 20, HEBDNA-binding protein HTF4, helix-loop-helix transcription factor 4, HsT17266, HTF4TCF-12, transcription factor 12, Transcription factor HTF-4 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human TCF12. The immunogen is located within amino acids 50 - 100 of TCF12. Amino Acid Squence: QPSPSYDSSRGFTDSPHYS |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?