missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCERG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35474-100ul
This item is not returnable.
View return policy
Description
TCERG1 Polyclonal antibody specifically detects TCERG1 in Human samples. It is validated for ELISA,Western Blot
Specifications
| TCERG1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| CA150TATA box-binding protein-associated factor 2S, co-activator of 150 kDa, TAF2SMGC133200, TATA box binding protein (TBP)-associated factor, RNA polymerase II, S, 150kD, TATA box-binding protein-associated factor 2S, transcription elongation regulator 1, Transcription factor CA150, Urn1 | |
| A synthetic peptide corresponding to a sequence within amino acids 550-650 of human TCERG1 (NP_006697.2).,, Sequence:, TTRLSMWDRPDDLIGRADVDKIIQEPPHKKGMEELKKLRHPTPTMLSIQKWQFSMSAIKEEQELMEEINEDEPVKAKKRKRDDNKDIDSEKEAAMEAEIKA | |
| 100 μL | |
| Cellular Markers, Transcription Factors and Regulators | |
| 10915 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction