missing translation for 'onlineSavingsMsg'
Learn More

TCEP Hydrochloride (0.5M in water), TRC

Product Code. 30412912
Change view
Click to view available options
Quantity:
1 mL
10 mL
Unit Size:
10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412912 1 mL -
30415185 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30412912 Supplier Toronto Research Chemicals Supplier No. TRCT0125151ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51805-45-9
Molecular Formula C9 H15 O6 P . Cl H
Molecular Weight (g/mol) 286.65
Synonym Tris (2-Carboxyethyl) Phosphine Hydrochloride (TCEP)
IUPAC Name 3-[bis(2-carboxyethyl)phosphanyl]propanoic acid;hydrochloride
SMILES Cl.OC(=O)CCP(CCC(=O)O)CCC(=O)O

Specifications

Quantity 1 mL
InChI Formula InChI=1S/C9H15O6P.ClH/c10-7(11)1-4-16(5-2-8(12)13)6-3-9(14)15;/h1-6H2,(H,10,11)(H,12,13)(H,14,15);1H
Formula Weight 286.0373
Concentration or Composition (by Analyte or Components) 0.5 M
Chemical Name or Material TCEP Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.