missing translation for 'onlineSavingsMsg'
Learn More

TCEB3B Antibody, Novus Biologicals™

Product Code. 18327599 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327599 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327599 Supplier Novus Biologicals Supplier No. H00051224B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

TCEB3B Polyclonal antibody specifically detects TCEB3B in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCEB3B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. AAI03912.1
Gene Alias 110kDa Elongin A, EloA2, Elongin-A2, HsT832, MGC119351, RNA polymerase II transcription factor SIII subunit A2, TCEB3Ltranscription elongation factor (SIII) elongin A2 elongin A2, Transcription elongation factor B polypeptide 3B, transcription elongation factor B polypeptide 3B (elongin A2)
Host Species Mouse
Immunogen TCEB3B (AAI03912.1, 1 a.a. - 753 a.a.) full-length human protein. MAAGSTTLHAVEKLQVRLATKTEPKKLEKYLQKLSALLMTADILAETGIRKTVKRLRKHQHVGDFARDLAARWKKLVLVDRNTRPGPQDPEESASRQRFGEALQDQEKAWGFPENATAPRSPSHSPEHRRTARRTPPGQQRPHPRSHSREPRAERKCPRIAPADSGRYRASPTRTAPLPMPEGPEPAAPGKQPGRGHTHAAQGGPLLCPGCQGQPQGKAVVSHSKGHKSSRQEKRPLCAQGDWHSPTLIREKSFGACLREETPRMPSWASARDRQPSDFKTDKEGGQAGSGQRVPALEEAPDSHQKRPQHSHSNKKRPSLDGRDPGNGTHGLSPEEKEQLSNDRETQEGKPPTAHLDRTSVSSLSEVEEVDMAEEFEQPTLSCEKYLTYDQLRKQKKKTGKSSTTALGDKQRKANESKGTRESWDSAKKLPPVQESQSERLQAAGTDSAGPKTVPSHVFSELWDLSEAWMQANYDPLSDSDSMTSQAKPEALSSPKFREEAAFPGRRVNAKMPVYSGSRPACQLQVPTLRQQCAQVLRNNPDALSDVGEVPYWVLEPVLEGWRPDQLYRRKKDNHALVRETDELRRNHCFQDFKEEKPQENKTWREQYLRLPDAPEQRLRVMTTNIRSARGNNPNGREAKMICFKSVAKTPYDTSRRQEKSAGDADPENGEIKPASKPAGSSHTPSSQSSSGGGRDSSSSILRWLPEKRANPCLSSSNEHAAPAAKTRKQAAKKVAPLMAKAIRDYKGRFSRR
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51224
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.