missing translation for 'onlineSavingsMsg'
Learn More

TCEB3B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-38555-20ul

 View more versions of this product

Product Code. 30227517

  • £145.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TCEB3B Polyclonal antibody specifically detects TCEB3B in Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TCEB3B
Polyclonal
Western Blot 1:500 - 1:2000, ELISA
110kDa Elongin A, EloA2, Elongin-A2, HsT832, MGC119351, RNA polymerase II transcription factor SIII subunit A2, TCEB3Ltranscription elongation factor (SIII) elongin A2 elongin A2, Transcription elongation factor B polypeptide 3B, transcription elongation factor B polypeptide 3B (elongin A2)
Recombinant fusion protein containing a sequence corresponding to amino acids 534-753 of human TCEB3B (NP_057511.2).,, Sequence:, AQVLRNNPDALSDVGEVPYWVLEPVLEGWRPDQLYRRKKDNHALVRETDELRRNHCFQDFKEEKPQENKTWREQYLRLPDAPEQRLRVMTTNIRSARGNNPNGREAKMICFKSVAKTPYDTSRRQEKSAGDADPENGEIKPASKPAGSSHTPSSQSSSGGGRDSSSSILRWLPEKRANPCLSSSNEHAAPAAKTRKQAAKKVAPLMAKAIRDYKRRFSRR
20 μL
Primary
Mouse, Rat
Purified
ELISA, Western Blot
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
51224
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.