missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEB2 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33277-100ul
This item is not returnable.
View return policy
Description
TCEB2 Monoclonal antibody specifically detects TCEB2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| TCEB2 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| 18-kD subunit, EloB, Elongin 18 kDa subunit, elongin-B, RNA polymerase II transcription factor SIII p18 subunit, RNA polymerase II transcription factor SIII subunit B, SIII p18, transcription elongation factor B (SIII), polypeptide 2 (18kD, elongin B), transcription elongation factor B (SIII), polypeptide 2 (18kDa, elongin B), transcription elongation factor B polypeptide 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human TCEB2 (NP_009039.1).,, Sequence:, MDVFLMIRRHKTTIFTDAKESSTVFELKRIVEGILKRPPDEQRLYKDDQLLDDGKTLGECGFTSQTARPQAPATVGLAFRADDTFEALCIEPFSSPPELP | |
| 100 μL | |
| Cell Biology | |
| 6923 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction