missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEB1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | TCEB1 |
|---|---|
| Clone | 2D6D8 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
30233542
|
Proteintech
68164-1-IG-150UL |
150 μL |
£349.00 £295.00 / 150µL Save £54.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234077
|
Proteintech
68164-1-IG-20UL |
20 μL |
£135.00 £119.00 / 20µL Save £16.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
The TCEB1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TCEB1. This antibody recognizes human, mouse, rat antigen. The TCEB1 antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Spezifikation
| TCEB1 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 6921 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 2D6D8 | |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| SIII | |
| IgG1 | |
| Protein A purification | |
| Monoclonal Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts