missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEANC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£342.00 - £470.00
Specifications
| Antigen | TCEANC |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18419040
|
Novus Biologicals
NBP1-81184-25ul |
25ul |
£342.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18246658
|
Novus Biologicals
NBP1-81184 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCEANC Polyclonal specifically detects TCEANC in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TCEANC | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 170082 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ETDVVRAVYRVLKNCPSVALKKKAKCLLSKWKAVYKQTHSKARNSPKLFPVRGNKEENSGPSHDPSQNETLGICSSNSLSSQDVAKLSEMIVPENRAIQLKPKEEHFGDGDPESTGK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| MGC17403, TFIIS central domain-containing protein 1, TFS2-M domain-containing protein 1, transcription elongation factor A (SII) N-terminal and central domaincontaining, transcription elongation factor A N-terminal and central domain-containingprotein | |
| TCEANC | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title