missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCEAL4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550145
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total TCEAL4 protein.
May be involved in transcriptional regulation. Lu X.W., Zhongguo Yi Xue Ke Xue Yuan Xue Bao 23:27-31(2001). Xu X., Submitted (MAY-2000) to the EMBL/GenBank/DDBJ databases. Ota T., Nat. Genet. 36:40-45(2004).
Specifications
| TCEAL4 | |
| Polyclonal | |
| Unconjugated | |
| TCEAL4 | |
| NPD017; TCEAL4; TCEA-like protein 4; transcription elongation factor A (SII)-like 4; transcription elongation factor A like 4; transcription elongation factor A protein-like 4; Transcription elongation factor S-II protein-like 4; WEX7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79921 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96EI5 | |
| TCEAL4 | |
| Synthesized peptide derived from internal of human TCEAL4 (aa 166-215). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction