missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCEAL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589056
This item is not returnable.
View return policy
Description
Immunogen sequence: MEKLFNENEG MPSNQGKIDN EEQPPHEGKP EVACILEDKK LENEGNTENT GKRVEEPLKD KEKPESAGKA KGEGKSERKG KSEMQGGSKT EGKPERGGRA EGEGEPDSER; Positive Samples: Mouse brain; Cellular Location: Nucleus.
TCEAL2 is a member of the transcription elongation factor A (SII)-like (TCEAL) gene family. Members of this family contain TFA domains and may function as nuclear phosphoproteins that modulate transcription in a promoter context-dependent manner.This gene encodes a member of the transcription elongation factor A (SII)-like (TCEAL) gene family. Members of this family contain TFA domains and may function as nuclear phosphoproteins that modulate transcription in a promoter context-dependent manner. Multiple family members are located on the X chromosome.
Specifications
| TCEAL2 | |
| Polyclonal | |
| Unconjugated | |
| my048; MY0876G0; MY0876G05; TCEAL2; TCEA-like protein 2; transcription elongation factor A (SII)-like 2; transcription elongation factor A like 2; transcription elongation factor A protein-like 2; Transcription elongation factor S-II protein-like 2; WEX1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human TCEAL2 (NP_5251291). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.68 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction