missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCEAL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567419
This item is not returnable.
View return policy
Description
Immunogen sequence: KTEGKPERGGR AEGEGEPDSE REPE Highest antigen sequence identity to the following orthologs - mouse 54%, rat 54%.
TCEAL2 is a member of the transcription elongation factor A (SII)-like (TCEAL) gene family. Members of this family contain TFA domains and may function as nuclear phosphoproteins that modulate transcription in a promoter context-dependent manner.This gene encodes a member of the transcription elongation factor A (SII)-like (TCEAL) gene family. Members of this family contain TFA domains and may function as nuclear phosphoproteins that modulate transcription in a promoter context-dependent manner. Multiple family members are located on the X chromosome.
Specifications
| TCEAL2 | |
| Polyclonal | |
| Unconjugated | |
| TCEAL2 | |
| my048; MY0876G0; MY0876G05; TCEAL2; TCEA-like protein 2; transcription elongation factor A (SII)-like 2; transcription elongation factor A like 2; transcription elongation factor A protein-like 2; Transcription elongation factor S-II protein-like 2; WEX1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140597 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H3H9 | |
| TCEAL2 | |
| Recombinant Human TCEAL2. Recombinant protein control fragment (Product #RP-108082). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction