missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEAL1 Antibody (OTI2G10), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74472UV
This item is not returnable.
View return policy
Description
TCEAL1 Monoclonal specifically detects TCEAL1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TCEAL1 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TCEAL1 | |
| Full length recombinant protein of human TCEAL1 (NP_004771) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2G10 | |
| Western Blot, Immunohistochemistry | |
| Nuclear phosphoprotein p21/SIIR, p21, pp21, SIIRTCEA-like protein 1, transcription elongation factor A (SII)-like 1, transcription elongation factor A protein-like 1, Transcription elongation factor S-II protein-like 1 | |
| Mouse | |
| Affinity Purified | |
| DNA replication Transcription Translation and Splicing | |
| 9338.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction