missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCEA3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560872
This item is not returnable.
View return policy
Description
Immunogen sequence: AESPKTPSSP LTPTFASSMC LLAPCYLTGD SVRDKCVEML SAALKADDDY KDYGVNCDKM ASEIEDH Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.
Necessary for efficient RNA polymerase II transcription elongation past template-encoded arresting sites. The arresting sites in DNA have the property of trapping a certain fraction of elongating RNA polymerases that pass through, resulting in locked ternary complexes. Cleavage of the nascent transcript by S-II allows the resumption of elongation from the new 3'-terminus.
Specifications
| TCEA3 | |
| Polyclonal | |
| Unconjugated | |
| TCEA3 | |
| hypothetical protein LOC402983; MGC137536 protein; rhabdomyosarcoma antigen MU-RMS-40.22; S-II; SII-K1; Tcea3; TFIIS; TFIIS.H; Tfiish; tissue-specific transcription factor S-II; transcription elongation factor A (SII), 3; transcription elongation factor A protein 3; transcription elongation factor A3; transcription elongation factor S-II protein 3; Transcription elongation factor TFIIS.h; zgc:77732 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6920 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75764 | |
| TCEA3 | |
| Recombinant protein corresponding to Human TCEA3. Recombinant protein control fragment (Product #RP-94579). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction