missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TCEA1 Recombinant Rabbit Monoclonal Antibody (K01_2J13)

Product Code. 30828214
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828214 50 μL
30827118 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828214 Supplier Invitrogen™ Supplier No. MA565073
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

TCEA1 Recombinant Monoclonal Antibody for Western Blot

TCEA1 is necessary for efficient RNA polymerase II transcription elongation past template-encoded arresting sites. TCEA1 has a role in suppression of transient pausing, which is the most important contribution of TCEA1 to elongation from a stall position. TCEA1 fuse to PLAG1 protein which is overexpressed in epithelial, myoepithelial, and mesenchymal-like tumor cells in tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCEA1
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2J13
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene TCEA1
Gene Accession No. P23193, Q4KLL0
Gene Alias GTF2S; SII; S-II; TCEA; Tcea1; Tceat; TF2S; TFIIS; transcription elongation factor A (SII) 1; transcription elongation factor A (SII), 1; transcription elongation factor A protein 1; Transcription elongation factor A protein 1-like protein; transcription elongation factor A1; transcription elongation factor S-II protein 1; Transcription elongation factor TFIIS.o; transcription factor IIS
Gene Symbols TCEA1
Host Species Rabbit
Immunogen A synthetic peptide of human TCEA1.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362479, 6917
Target Species Human, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.