missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEA1 Recombinant Rabbit Monoclonal Antibody (23GB2330), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA552502
This item is not returnable.
View return policy
Description
TCEA1 Recombinant Monoclonal antibody specifically detects TCEA1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications
| TCEA1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| TCEA1 | |
| GTF2S; SII; S-II; TCEA; Tcea1; Tceat; TF2S; TFIIS; transcription elongation factor A (SII) 1; transcription elongation factor A (SII), 1; transcription elongation factor A protein 1; Transcription elongation factor A protein 1-like protein; transcription elongation factor A1; transcription elongation factor S-II protein 1; Transcription elongation factor TFIIS.o; transcription factor IIS | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 21399, 362479, 6917 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2330 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P10711, P23193, Q4KLL0 | |
| TCEA1 | |
| A synthesized peptide derived from human TCEA1 (110-134AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction