missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEA1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94175-0.1ml
This item is not returnable.
View return policy
Description
TCEA1 Polyclonal antibody specifically detects TCEA1 in Human samples. It is validated for Western Blot
Specifications
| TCEA1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| GTF2Stranscription elongation factor A protein 1, SII, TF2STCEA, TFIISTranscription elongation factor TFIIS.o, transcription elongation factor A (SII), 1, Transcription elongation factor S-II protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 75-140 of human TCEA1 (NP_006747.1). KLLDGPSTEKDLDEKKKEPAITSQNSPEAREESTSSGNVSNRKDETNARDTYVSSFPRAPSTSDSV | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 6917 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction