missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TCB-2
GREENER_CHOICE

Product Code. p-7112877
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18064212

Brand: Tocris Bioscience™ 2592/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity and potent 5-HT2A agonist

TCB-2 is a high affinity 5-HT2A receptor agonist (Ki values are 0.73 and 0.75 nM for rat and human receptors respectively). Potently stimulates IP3 accumulation in NIH3T3 cells stably expressing rat 5-HT2A receptors (EC50 = 36 nM). Induces head twitches and hypothermia in mice following i.p. administration.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (4-Bromo-3,6-dimethoxybenzocyclobuten-1-yl)methylamine hydrobromide
CAS 912342-28-0
Assay Percent Range >99%
Quantity 50 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C11H14BrNO2.HBr
Formula Weight Observed MW: 353.05

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.