missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCAP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555315
This item is not returnable.
View return policy
Description
Immunogen sequence: CEVSEENCER REAFWAEWKD LTLSTRPEEG CSLHEEDTQR HETYHQQGQC QVLVQRSPWL MMRMGILGRG LPEYQ Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 65%.
Sarcomere assembly is regulated by the muscle protein titin. Titin is a giant elastic protein with kinase activity that extends half the length of a sarcomere. It serves as a scaffold to which myofibrils and other muscle related proteins are attached. TCAP is a protein found in striated and cardiac muscle that binds to the titin Z1-Z2 domains and is a substrate of titin kinase, interactions thought to be critical to sarcomere assembly. Mutations in TCAP gene are associated with limb-girdle muscular dystrophy type 2G.Sarcomere assembly is regulated by the muscle protein titin. Titin is a giant elastic protein with kinase activity that extends half the length of a sarcomere. It serves as a scaffold to which myofibrils and other muscle related proteins are attached. This gene encodes a protein found in striated and cardiac muscle that binds to the titin Z1-Z2 domains and is a substrate of titin kinase, interactions thought to be critical to sarcomere assembly. Mutations in this gene are associated with limb-girdle muscular dystrophy type 2G.
Specifications
| TCAP | |
| Polyclonal | |
| Unconjugated | |
| Tcap | |
| 19 kDa sarcomeric protein; AI503981; AJ223855; CMD1N; CMH25; LGMD2G; limb girdle muscular dystrophy 2G (autosomal recessive); Tcap; T-cap; TELE; telethonin; teneurin C-terminal associated peptide; titin cap protein; titin-cap; titin-cap (telethonin) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8557 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15273 | |
| Tcap | |
| Recombinant protein corresponding to Human TCAP. Recombinant protein control fragment (Product #RP-93427). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction