missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCAP Antibody (OTI11H10), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74471JF549
This item is not returnable.
View return policy
Description
TCAP Monoclonal specifically detects TCAP in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TCAP | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TCAP | |
| Full length recombinant protein of human TCAP (NP_003664) produced in E.coli. | |
| 0.1 mL | |
| 8557.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI11H10 | |
| Western Blot, Immunohistochemistry | |
| 19 kDa sarcomeric protein, CMD1NTitin cap protein, LGMD2G, limb girdle muscular dystrophy 2G (autosomal recessive), T-cap, TELE, telethonin, titin-cap (telethonin) | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction