missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TC21/R-Ras2 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81761H
This item is not returnable.
View return policy
Description
TC21/R-Ras2 Polyclonal antibody specifically detects TC21/R-Ras2 in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
| TC21/R-Ras2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| HRP | |
| ras-related protein R-Ras2, related RAS viral (r-ras) oncogene homolog 2, TC21Ras-like protein TC21, Teratocarcinoma oncogene | |
| Rabbit polyclonal R-Ras2 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human R-Ras2. The immunogen is located within the last 50 amino acids of R-Ras2. Amino Acid Squence: PPSPEPTRKEKDKKG | |
| 0.1 mL | |
| Signal Transduction | |
| 22800 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction