missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TC21/R-Ras2 Antibody (EM-50), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62216PE
This item is not returnable.
View return policy
Description
TC21/R-Ras2 Monoclonal specifically detects TC21/R-Ras2 in Human, Monkey samples. It is validated for Flow Cytometry, Flow (Intracellular).
Specifications
| TC21/R-Ras2 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| RRAS2 | |
| Human TC21 | |
| 0.1 mL | |
| Primary | |
| The antibody MEM-31 recognizes a conformationally-dependent epitope of CD8, a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediates efficient cell-cell interactions within the immune system. CD8 is a disulfide-linked dimer and exists as a CD8 alpha/alpha homodimer or CD8 alpha/beta heterodimer (each monomer approx. 32-34 kDa). The antibody does not react with formaldehyde-fixed cells; negative in Western Blotting application. | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| EM-50 | |
| Flow Cytometry, Flow (Intracellular) | |
| ras-related protein R-Ras2, related RAS viral (r-ras) oncogene homolog 2, TC21Ras-like protein TC21, Teratocarcinoma oncogene | |
| Mouse | |
| Protein A purified | |
| Signal Transduction | |
| 22800 | |
| Human, Monkey | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction