missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TC21/R-Ras2 Monoclonal antibody specifically detects TC21/R-Ras2 in Human, Monkey samples. It is validated for Flow Cytometry, Flow (Intracellular)
Specifications
Specifications
| Antigen | TC21/R-Ras2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | EM-50 |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Flow Cytometry, Flow (Intracellular) |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ras-related protein R-Ras2, related RAS viral (r-ras) oncogene homolog 2, TC21Ras-like protein TC21, Teratocarcinoma oncogene |
| Host Species | Mouse |
| Immunogen | Human TC21 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?