missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TC-S 7010

Product Code. p-7110802
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18711829

Brand: Tocris Bioscience™ 5286/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent and selective Aurora kinase A inhibitor

TC-S 7010 is a potent and selective inhibitor of Aurora kinase A (IC50 = 3.4 nM); exhibits 1000-fold selectivity for Aurora kinase A over Aurora kinase B. Displays antiproliferative activity in HCT116 and HT29 cells (IC50 values are 0.19 and 2.9 μM respectively).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2-Chlorophenyl)-4-[[2-[[4-[2-(4-ethyl-1-piperazinyl)-2-oxoethyl]phenyl]amino]-5-fluoro-4-pyrimidinyl]amino]-benzamide
CAS 1158838-45-9
Quantity 10 mg
Target Aurora Kinase Inhibitors
Molecular Formula C31H31ClFN7O2
Formula Weight 588.07
Purity >99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.