missing translation for 'onlineSavingsMsg'
Learn More

TC-S 7005, MedChemExpress

Product Code. 16476927
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476927 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16476927 Supplier Medchem Express Supplier No. HY108597
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress TC-S 7005 is a Polo-like kinases (Plks) inhibitor with IC50s of 4 nM, 24 nM and 214 nM for Plk2, Plk3, and Plk1, respectively.

TC-S 7005 is a Polo-like kinases (Plks) inhibitor with IC50s of 4 nM, 24 nM and 214 nM for Plk2, Plk3, and Plk1, respectively.

Specifications

Chemical Name or Material TC-S 7005
CAS 1082739-92-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H17N3O3
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (347.82 mM; Need ultrasonic)
SMILES C[C@H](NC1=CC2=C(ON=C2C3=CC=C(OCO4)C4=C3)C=N1)C5=CC=CC=C5
Molecular Weight (g/mol) 359.38
Formula Weight 359.38
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.