missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TC-S 7004

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
TC-S 7004 is a potent and selective DYRK1A/B inhibitor (IC50 values are 22 and 68 nM respectively). Selective for DYRK1A/B over a panel of other kinases. Reduces accumulation of quiescent Panc1 pancreatic cancer cells in G0 under serum free conditions. Also induces DNA damage and apoptosis in quiescent Panc1 cells.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | TC-S 7004 |
| Purity | >98% |
| Molecular Weight (g/mol) | 498.32 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction