missing translation for 'onlineSavingsMsg'
Learn More

TC-PTP/PTPN2 Antibody (001), DyLight 680, Novus Biologicals™

Product Code. 18387120 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18387120 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18387120 Supplier Novus Biologicals Supplier No. NBP290722FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TC-PTP/PTPN2 Monoclonal specifically detects TC-PTP/PTPN2 in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TC-PTP/PTPN2
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate DyLight 680
Formulation 50 mM Sodium Borate
Gene Alias protein tyrosine phosphatase, non-receptor type 2, PTPTPTN2, T-cell protein tyrosine phosphatase, T-cell protein-tyrosine phosphatase, TCELLPTP, TC-PTP, TCPTPEC 3.1.3.48, tyrosine-protein phosphatase non-receptor type 2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TC-PTP/PTPN2 (Q06180-1; Ser2-Asn314).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5771
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.